Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMILIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018057881.1 | XP_017913370.1 | 3 - 4 | 515 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |