Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018063405.1 | XP_017918894.1 | 8 - 9 | 2477 | 67 | |
XM_018063406.1 | XP_017918895.1 | 8 - 9 | 2477 | 67 | ||
XM_018063407.1 | XP_017918896.1 | 9 - 10 | 1296 | 67 | ||
XM_018063408.1 | XP_017918897.1 | 8 - 9 | 1233 | 67 | ||
XM_018063409.1 | XP_017918898.1 | 8 - 9 | 923 | 67 | ||
XM_018063410.1 | XP_017918899.1 | 2 - 3 | 2116 | 67 | ||
GenBank mRNA | JO426493.1 | - | 8 - 9 | 1007 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |