Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP6D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005675264.3 | XP_005675321.2 | 1 - 2 | 437 | 68 | |
GenBank mRNA | JO641367.2 | - | 1 - 2 | 410 | 68 | |
KA333364.1 | - | 2 - 3 | 302 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |