Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP6D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005675264.3 | XP_005675321.2 | 2 - 3 | 557 | 77 | |
GenBank mRNA | JO641367.2 | - | 2 - 3 | 530 | 77 | |
KA333364.1 | - | 3 - 4 | 422 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |