Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005682840.2 | XP_005682897.2 | 5 - 6 | 875 | 61 | |
GenBank mRNA | JR121379.1 | - | 716 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |