Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AZU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005682894.3 | XP_005682951.1 | 2 - 3 | 507 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Capra hircus Probe spans exons |