Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBLN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013966815.2 | XP_013822269.1 | 2 - 3 | 1198 | 64 | |
XM_013966816.2 | XP_013822270.1 | 2 - 3 | 1198 | 64 | ||
XM_013966818.2 | XP_013822272.1 | 2 - 3 | 1198 | 64 | ||
XM_013966819.2 | XP_013822273.1 | 2 - 3 | 1198 | 64 | ||
XM_018054655.1 | XP_017910144.1 | 2 - 3 | 1198 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |