Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOC3L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018062588.1 | XP_017918077.1 | 9 - 10 | 1218 | 68 | |
XM_018062589.1 | XP_017918078.1 | 8 - 9 | 1049 | 68 | ||
GenBank mRNA | JO428051.1 | - | 11 - 12 | 2268 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Capra hircus Probe spans exons |