Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM71F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005679438.3 | XP_005679495.1 | 1 - 2 | 252 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |