Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005679525.3 | XP_005679582.2 | 2 - 3 | 395 | 76 | |
XM_018046866.1 | XP_017902355.1 | 2 - 3 | 307 | 76 | ||
XM_018046867.1 | XP_017902356.1 | 2 - 3 | 407 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |