Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018054095.1 | XP_017909584.1 | 3 - 4 | 1841 | 64 | |
XM_018054096.1 | XP_017909585.1 | 3 - 4 | 1842 | 64 | ||
GenBank mRNA | KA315308.1 | - | 5 - 6 | 1383 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |