Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005687480.3 | XP_005687537.2 | 2 - 3 | 593 | 82 | |
GenBank mRNA | JO437335.2 | - | 5 - 6 | 785 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |