Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMED6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692188.3 | XP_005692245.1 | 1 - 2 | 585 | 66 | |
XM_018062211.1 | XP_017917700.1 | 1 - 2 | 1368 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |