Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005700785.3 | XP_005700842.1 | 5 - 6 | 566 | 93 | |
XM_005700786.3 | XP_005700843.1 | 5 - 6 | 567 | 93 | ||
XM_013976462.2 | XP_013831916.1 | 5 - 6 | 594 | 93 | ||
GenBank mRNA | JO642884.2 | - | 2 - 3 | 207 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Capra hircus Probe spans exons |