Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018046711.1 | XP_017902200.1 | 4 - 5 | 1391 | 58 | |
XM_018046712.1 | XP_017902201.1 | 4 - 5 | 1225 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Capra hircus Probe spans exons |