Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102188408 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005685214.3 | XP_005685271.1 | 2 - 3 | 575 | 70 | |
XM_005685215.3 | XP_005685272.1 | 2 - 3 | 558 | 70 | ||
GenBank mRNA | JO450440.2 | - | 2 - 3 | 197 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Capra hircus Probe spans exons |