Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCLAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055477.1 | XP_017910966.1 | 2 - 3 | 321 | 89 | |
GenBank mRNA | JT762624.1 | - | 1 - 2 | 70 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |