Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102191731 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693058.2 | XP_005693115.2 | 1 - 2 | 401 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |