Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM241B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042627.1 | XP_017898116.1 | 2 - 3 | 395 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |