Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7H1orf35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051585.1 | XP_017907074.1 | 7 - 8 | 963 | 145 | |
XM_018051586.1 | XP_017907075.1 | 6 - 7 | 761 | 145 | ||
GenBank mRNA | KA308604.1 | - | 6 - 7 | 867 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Capra hircus Probe spans exons |