Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064568.1 | XP_017920057.1 | 2 - 3 | 414 | 91 | |
GenBank mRNA | KP408266.1 | - | 2 - 3 | 284 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |