Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STMN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005676827.3 | XP_005676884.1 | 3 - 4 | 267 | 107 | |
XM_018056181.1 | XP_017911670.1 | 3 - 4 | 355 | 107 | ||
GenBank mRNA | JO339694.2 | - | 2 - 3 | 741 | 107 | |
KA335298.1 | - | 2 - 3 | 716 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Capra hircus Probe spans exons |