Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3H1orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677681.3 | XP_005677738.1 | 2 - 3 | 737 | 127 | |
XM_005677682.3 | XP_005677739.1 | 2 - 3 | 737 | 127 | ||
GenBank mRNA | JO578970.1 | - | 4 - 5 | 416 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |