Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3H1orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677681.3 | XP_005677738.1 | 4 - 5 | 905 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |