Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C26H10orf82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041132.1 | XP_017896621.1 | 4 - 5 | 563 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |