Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042529.1 | XP_017898018.1 | 2 - 3 | 823 | 76 | |
GenBank mRNA | KA316115.1 | - | 2 - 3 | 701 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |