Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042529.1 | XP_017898018.1 | 5 - 6 | 1195 | 84 | |
GenBank mRNA | JR131735.1 | - | 1952 | 84 | ||
KA316115.1 | - | 5 - 6 | 1073 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |