Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RALY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018057731.1 | XP_017913220.1 | 6 - 7 | 1097 | 88 | |
XM_018057732.1 | XP_017913221.1 | 5 - 6 | 1014 | 88 | ||
XM_018057733.1 | XP_017913222.1 | 5 - 6 | 1049 | 88 | ||
GenBank mRNA | JT682410.1 | - | 7 - 8 | 1164 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |