Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDBF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018066546.1 | XP_017922035.1 | 1 - 2 | 58 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |