Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WFDC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005688586.3 | XP_005688643.1 | 1 - 2 | 172 | 61 | |
GenBank mRNA | KA326283.1 | - | 1 - 2 | 101 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |