Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTP4A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058643.1 | XP_017914132.1 | 2 - 3 | 1066 | 66 | |
XM_018058644.1 | XP_017914133.1 | 2 - 3 | 990 | 66 | ||
XM_018058645.1 | XP_017914134.1 | 2 - 3 | 958 | 66 | ||
GenBank mRNA | JO363644.2 | - | 945 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |