Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STX1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041038.1 | XP_017896527.1 | 4 - 5 | 338 | 57 | |
XM_018041039.1 | XP_017896528.1 | 4 - 5 | 331 | 57 | ||
GenBank mRNA | JO626976.2 | - | 6 - 7 | 697 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Capra hircus Probe spans exons |