Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FADS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018063849.1 | XP_017919338.1 | 4 - 5 | 795 | 102 | |
GenBank mRNA | JO421557.2 | - | 2 - 3 | 1882 | 102 | |
KA317111.1 | - | 2 - 3 | 2664 | 102 | ||
KT315580.1 | - | 4 - 5 | 743 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |