Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043072.1 | XP_017898561.1 | 4 - 5 | 1310 | 75 | |
GenBank mRNA | KA344435.1 | - | 905 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |