Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058059.1 | XP_017913548.1 | 3 - 4 | 450 | 55 | |
XM_018058061.1 | XP_017913550.1 | 3 - 4 | 450 | 55 | ||
GenBank mRNA | KA348401.1 | - | 3 - 4 | 1100 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |