Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005696417.3 | XP_005696474.2 | 5 - 6 | 695 | 98 | |
XM_005696421.3 | XP_005696478.2 | 4 - 5 | 672 | 98 | ||
XM_013973902.2 | XP_013829356.2 | 6 - 7 | 1013 | 98 | ||
XM_013973903.2 | XP_013829357.2 | 6 - 7 | 1075 | 98 | ||
XM_013973904.2 | XP_013829358.2 | 6 - 7 | 718 | 98 | ||
XM_013973906.2 | XP_013829360.2 | 4 - 5 | 485 | 98 | ||
XM_018039201.1 | XP_017894690.1 | 6 - 7 | 1092 | 98 | ||
XM_018039202.1 | XP_017894691.1 | 6 - 7 | 856 | 98 | ||
XM_018039203.1 | XP_017894692.1 | 6 - 7 | 978 | 98 | ||
XM_018039204.1 | XP_017894693.1 | 6 - 7 | 847 | 98 | ||
XM_018039205.1 | XP_017894694.1 | 5 - 6 | 718 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |