Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCTPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041031.1 | XP_017896520.1 | 1 - 2 | 200 | 59 | |
GenBank mRNA | JO587331.2 | - | 1 - 2 | 150 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |