Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCAMP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018050406.1 | XP_017905895.1 | 6 - 7 | 650 | 101 | |
XM_018050407.1 | XP_017905896.1 | 6 - 7 | 617 | 101 | ||
GenBank mRNA | JO632047.2 | - | 6 - 7 | 643 | 101 | |
KA332497.1 | - | 1 - 2 | 1537 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Capra hircus Probe spans exons |