Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013966433.2 | XP_013821887.2 | 1 - 2 | 1285 | 89 | |
XM_018053071.1 | XP_017908560.1 | 2 - 3 | 619 | 89 | ||
XM_018053072.1 | XP_017908561.1 | 1 - 2 | 1294 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |