Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064074.1 | XP_017919563.1 | 2 - 3 | 861 | 104 | |
XM_018064075.1 | XP_017919564.1 | 2 - 3 | 350 | 104 | ||
GenBank mRNA | JO557133.2 | - | 3 - 4 | 565 | 104 | |
JO645222.2 | - | 2 - 3 | 1672 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |