Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNGA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058978.1 | XP_017914467.1 | 2 - 3 | 275 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |