Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASEK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693473.3 | XP_005693530.1 | 1 - 2 | 203 | 93 | |
GenBank mRNA | JO568344.2 | - | 2 - 3 | 480 | 93 | |
JO612686.2 | - | 2 - 3 | 461 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |