Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C20H5orf22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005694829.3 | XP_005694886.1 | 3 - 4 | 454 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |