Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018052049.1 | XP_017907538.1 | 3 - 4 | 425 | 77 | |
GenBank mRNA | JO390910.2 | - | 3 - 4 | 421 | 77 | |
KA344975.1 | - | 2 - 3 | 1071 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |