Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2D6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055381.1 | XP_017910870.1 | 12 - 13 | 693 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |