Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005689986.2 | XP_005690043.1 | 2 - 3 | 220 | 67 | |
GenBank mRNA | JO580043.1 | - | 5 - 6 | 1112 | 67 | |
KA340194.1 | - | 2 - 3 | 208 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |