Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM83G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064758.1 | XP_017920247.1 | 4 - 5 | 2140 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |