Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM2D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018066496.1 | XP_017921985.1 | 5 - 6 | 626 | 106 | |
XM_018066497.1 | XP_017921986.1 | 4 - 5 | 533 | 106 | ||
GenBank mRNA | JO638732.2 | - | 5 - 6 | 651 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |