Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPY19L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005679195.2 | XP_005679252.1 | 13 - 14 | 1558 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |