Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDNF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005681260.3 | XP_005681317.2 | 2 - 3 | 599 | 87 | |
GenBank mRNA | JO506950.2 | - | 2 - 3 | 539 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |